siRNA Details
Gene Symbol: CT55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017863.2 | NP_060333.1 | 5 | 1614 | ||
GenBank mRNA | AK000534.1 | - | 5 | 1121 | ||
AY352211.1 | - | 5 | 1614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
