siRNA Details
Gene Symbol: C5orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016175.3 | NP_057259.2 | 3 | 230 | ||
GenBank mRNA | AK293901.1 | - | 3 | 196 | ||
BC069051.1 | - | 3 | 198 | 3945859 | ||
BF965313.1 | - | 3 | 231 | |||
BX537968.1 | - | 4 | 331 | |||
BX647375.1 | - | 3 | 198 | |||
CN363239.1 | - | 4 | 234 | |||
CR936785.1 | - | 3 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
