siRNA Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021647.7 | NP_067679.6 | 2 | 425 | ||
XM_005263366.3 | XP_005263423.1 | 3 | 359 | |||
XM_017008864.1 | XP_016864353.1 | 4 | 303 | |||
XM_017008865.1 | XP_016864354.1 | 4 | 3419 | |||
XM_017008866.1 | XP_016864355.1 | 4 | 493 | |||
XM_017008867.1 | XP_016864356.1 | 4 | 299 | |||
XM_017008868.1 | XP_016864357.1 | 3 | 574 | |||
XM_017008869.1 | XP_016864358.1 | 2 | 3406 | |||
XM_017008870.1 | XP_016864359.1 | 2 | 3366 | |||
XM_017008871.1 | XP_016864360.1 | 2 | 306 | |||
GenBank mRNA | AB014526.1 | - | 2 | 273 | ||
AK290041.1 | - | 2 | 335 | |||
AY391838.1 | - | 1 | 206 | |||
BC001279.1 | - | 3 | 713 | |||
DA324165.1 | - | 2 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
