siRNA Details
Gene Symbol: Airn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK035282.1 | - | 1 | 1678 | ||
AU015441.1 | - | LIT1853 Air |
1 | 100 | ||
BG070272.2 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
