siRNA Details
Gene Symbol: C20orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039140.2 | NP_001034229.1 | 4, 5 | 613 | ||
NM_001258429.1 | NP_001245358.1 | 4, 5 | 538 | |||
NM_001258430.1 | NP_001245359.1 | 4, 5 | 992 | |||
NR_047675.1 | 4, 5 | 682 | ||||
GenBank mRNA | AK000557.1 | - | 4, 5 | 477 | ||
AK024220.1 | - | 4, 5 | 477 | |||
AK097645.1 | - | 4, 5 | 1120 | |||
AK125202.1 | - | 2, 3 | 1446 | |||
AK225109.1 | - | 4, 5 | 477 | |||
AK290955.1 | - | 4, 5 | 682 | |||
BC012196.1 | - | 4, 5 | 464 | 2822218 | ||
BC024036.1 | - | 4, 5 | 475 | 3510659 | ||
BF978883.1 | - | 4, 5 | 539 | |||
BG474804.1 | - | 4, 5 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
