siRNA Details
Gene Symbol: LOC500227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024330.1 | NP_001019501.1 | 2 | 263 | ||
XM_006236784.2 | XP_006236846.1 | 2 | 272 | |||
GenBank mRNA | BC079424.1 | - | 2 | 263 | 7114113 |
