siRNA Details
Gene Symbol: C1orf123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304759.1 | NP_001291688.1 | 6, 7 | 420 | ||
NM_001304760.1 | NP_001291689.1 | 5, 6 | 336 | |||
NM_017887.2 | NP_060357.1 | 7, 8 | 477 | |||
GenBank mRNA | AK000587.1 | - | 7, 8 | 455 | ||
AK092734.1 | - | 5, 6 | 1321 | |||
AK094371.1 | - | 2, 3 | 1653 | |||
BC010908.1 | - | 7, 8 | 459 | 4093763 | ||
BE889129.1 | - | 4, 5 | 289 | |||
BI603716.1 | - | 6, 7 | 413 | |||
CR457246.1 | - | 6, 7 | 413 |
