siRNA Details
Gene Symbol: Fam110b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024341.1 | NP_001019512.1 | 1 | 1204 | ||
XM_006237839.2 | XP_006237901.1 | 3 | 2497 | |||
XM_006237840.2 | XP_006237902.1 | 4 | 1725 | |||
XM_006237841.2 | XP_006237903.1 | 3 | 1638 | |||
XM_006237843.2 | XP_006237905.1 | 2 | 6828 | |||
XM_008763551.1 | XP_008761773.1 | 2 | 7602 | |||
XM_008763552.1 | XP_008761774.1 | 3 | 5357 | |||
XM_008763553.1 | XP_008761775.1 | 4 | 5056 | |||
XM_008763554.1 | XP_008761776.1 | 4 | 5306 | |||
XM_008763555.1 | XP_008761777.1 | 2 | 2410 | |||
GenBank mRNA | BC091289.1 | - | 1 | 1204 | 7306431 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173426, AK159054, AK146011, BC055683, AK077232, and others.
