siRNA Details
Gene Symbol: LOC500475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024346.1 | NP_001019517.1 | 12 | 1510 | ||
XM_008763797.1 | XP_008762019.1 | 12 | 1753 | |||
XM_008763798.1 | XP_008762020.1 | 12 | 1293 | |||
XM_008763799.1 | XP_008762021.1 | 12 | 1103 | |||
GenBank mRNA | BC079053.1 | - | 12 | 1510 | 7115405 |
