siRNA Details
Gene Symbol: LOC500827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024352.1 | NP_001019523.2 | 9 | 1116 | ||
XM_006241290.2 | XP_006241352.1 | 9 | 982 | |||
XM_006241291.2 | XP_006241353.1 | 10 | 1614 | |||
XM_008765319.1 | XP_008763541.1 | 9 | 1635 | |||
GenBank mRNA | BC078911.1 | - | 9 | 1116 | 7112362 |
