siRNA Details
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY539942.1 | - | 2072 |
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY310153.1 | - | 861 | |||
AY318961.1 | - | 861 | ||||
AY321318.1 | - | 858 | ||||
AY321324.1 | - | 861 | ||||
AY325196.1 | - | 861 | ||||
AY325218.1 | - | 861 | ||||
AY325232.1 | - | 861 |
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY387051.1 | - | 2786 |
Gene Symbol: LOC100362110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S80119.1 | - | 835 |
Gene Symbol: LOC103694177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772007.1 | XP_008770229.1 | 35 |
Gene Symbol: LOC499182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ100474.1 | - | 3 | 4473 |
Gene Symbol: LOC499355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ100475.1 | - | 3 | 4470 |
Gene Symbol: LOC501503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ100477.1 | - | 2 | 4470 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U83119.1 | - | 1 | 2777 |

More Information
Other Species Targeted

