siRNA Details
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024354.1 | NP_001019525.1 | 4 | 1723 | ||
XM_006241860.2 | XP_006241922.1 | 4 | 1978 | |||
XM_006241861.1 | XP_006241923.1 | 4 | 1611 | |||
XM_006241862.2 | XP_006241924.1 | 5 | 2496 | |||
XM_006241865.1 | XP_006241927.1 | 5 | 1706 | |||
GenBank mRNA | BC092583.1 | - | 4 | 1723 | 7376401 |
