siRNA Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127229.1 | NP_001120701.1 | 905 | |||
NM_017900.2 | NP_060370.1 | 986 | ||||
GenBank mRNA | AK000615.1 | - | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
