siRNA Details
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025049.1 | NP_001020220.1 | 5 | 988 | ||
XM_006237379.2 | XP_006237441.1 | 5 | 1528 | |||
GenBank mRNA | BC079212.1 | - | 5 | 988 | 7131828 |
