siRNA Details
Gene Symbol: LOC500956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025054.1 | NP_001020225.1 | 7 | 1524 | ||
XM_006242662.2 | XP_006242724.1 | 8 | 1229 | |||
XM_008765978.1 | XP_008764200.1 | 8 | 1268 | |||
GenBank mRNA | BC086569.1 | - | 7 | 1524 | 7300711 |
