siRNA Details
Gene Symbol: Gmppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025056.1 | NP_001020227.1 | 13 | 1408 | ||
XM_006245267.2 | XP_006245329.1 | 14 | 1326 | |||
XM_006245268.2 | XP_006245330.1 | 14 | 1397 | |||
XM_006245269.2 | XP_006245331.1 | 14 | 1391 | |||
XM_006245270.2 | XP_006245332.1 | 14 | 1294 | |||
XM_006245271.2 | XP_006245333.1 | 13 | 1311 | |||
XM_008767291.1 | XP_008765513.1 | 14 | 1433 | |||
GenBank mRNA | BC083763.1 | - | 13 | 1408 | 7193128 |
