siRNA Details
Gene Symbol: LOC502684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025060.1 | NP_001020231.1 | 3 | 327 | ||
XM_006235246.2 | XP_006235308.1 | 3 | 368 | |||
XM_006235247.2 | XP_006235309.1 | 3 | 369 | |||
XM_006235248.2 | XP_006235310.1 | 3 | 467 | |||
XM_006235249.2 | XP_006235311.1 | 4 | 345 | |||
XM_006235251.2 | XP_006235313.1 | 3 | 367 | |||
XM_008762376.1 | XP_008760598.1 | 3 | 446 | |||
XM_008762377.1 | XP_008760599.1 | 4 | 405 | |||
XM_008762378.1 | XP_008760600.1 | 3 | 347 | |||
GenBank mRNA | BC083680.1 | - | 3 | 327 | 7135883 |
