siRNA Details
Gene Symbol: Ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025146.1 | NP_001020317.1 | 5 | 1354 | ||
GenBank mRNA | BC083729.1 | - | 5 | 1354 | 7191329 | |
BC127458.1 | - | 5 | 1354 | 7193742 | ||
FQ212751.1 | - | 5 | 1374 | |||
FQ214408.1 | - | 5 | 1371 |
