siRNA Details
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199308.2 | NP_955012.2 | 26 | 5320 | ||
GenBank mRNA | BC024265.1 | - | 9 | 3014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
