siRNA Details
Gene Symbol: Yipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199383.2 | NP_955415.1 | 9, 10 | 1063 | ||
GenBank mRNA | BC062239.1 | - | 9, 10 | 1067 | 6887537 | |
FQ218908.1 | - | 9, 10 | 1062 |
