siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 2770 | |||
GenBank mRNA | BC030620.1 | - | 2 | 2769 | ||
BC051704.1 | - | 2 | 2757 | |||
BP397928.1 | - | 1 | 366 | |||
CN430413.1 | - | 1 | 74 |
