siRNA Details
Gene Symbol: Olfr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206897.2 | NP_996780.2 | 1 | 14 | ||
GenBank mRNA | BC127998.1 | - | 1 | 18 | 40135788 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206897.2 | NP_996780.2 | 1 | 14 | ||
GenBank mRNA | BC127998.1 | - | 1 | 18 | 40135788 |