siRNA Details
Gene Symbol: LOC102724751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030164.1 | XP_016885653.1 | 2, 3 | 392 |
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 23 | 5454 | ||
NM_021574.2 | NP_067585.2 | 22 | 5322 | |||
GenBank mRNA | AK122842.1 | - | 6 | 2718 | ||
BC063619.1 | - | 1 | 84 | |||
BC083493.1 | - | 1 | 25 |
Gene Symbol: BCRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037566.1 | 5, 6 | 859 | |||
GenBank mRNA | AK124943.1 | - | 5, 6 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
