siRNA Details
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204817.1 | NP_001191746.1 | 3 | 4517 | ||
NM_032828.3 | NP_116217.1 | 3 | 4520 | |||
GenBank mRNA | AK000820.1 | - | 1 | 345 | ||
AK026676.1 | - | 1 | 774 | |||
AK160374.1 | - | 2 | 4396 | |||
BC017219.2 | - | 1 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
