siRNA Details
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517956.1 | XP_011516258.1 | 8 | 698 | ||
XM_011517957.1 | XP_011516259.1 | 5 | 590 | |||
XM_011517958.1 | XP_011516260.1 | 6 | 517 | |||
GenBank mRNA | AK000920.1 | - | 7 | 533 | ||
AK301790.1 | - | 8 | 798 | |||
BX649030.1 | - | 6 | 3179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
