siRNA Details
Gene Symbol: DPY19L2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002833.2 | 5 | 796 | |||
GenBank mRNA | AK304236.1 | - | 5 | 956 | ||
BC066987.1 | - | 5 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
