siRNA Details
Gene Symbol: TPCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143819.2 | NP_001137291.1 | 29 | 4601 | ||
NM_001301214.1 | NP_001288143.1 | 27 | 4183 | |||
NM_017901.5 | NP_060371.2 | 28 | 4508 | |||
XM_005253906.4 | XP_005253963.1 | 27 | 5022 | |||
XM_011538490.2 | XP_011536792.1 | 29 | 4567 | |||
XM_011538492.2 | XP_011536794.1 | 28 | 4414 | |||
XM_011538493.2 | XP_011536795.1 | 27 | 4258 | |||
XM_017019480.1 | XP_016874969.1 | 28 | 4473 | |||
XM_017019481.1 | XP_016874970.1 | 29 | 4613 | |||
GenBank mRNA | AB032995.2 | - | 28 | 4475 | ||
AK000619.1 | - | 1 | 871 | |||
AK025838.1 | - | 1 | 918 | |||
BC110885.1 | - | 1 | 973 | |||
BC150203.1 | - | 28 | 4431 | 40148827 | ||
BC152423.1 | - | 28 | 4475 | 8862535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
