siRNA Details
Gene Symbol: TMCO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017905.4 | NP_060375.4 | 3 | 954 | ||
XM_006719969.1 | XP_006720032.1 | 3 | 735 | |||
XM_011537498.2 | XP_011535800.1 | 3 | 1078 | |||
XM_011537501.2 | XP_011535803.1 | 3 | 954 | |||
XM_011537502.2 | XP_011535804.1 | 3 | 954 | |||
XM_011537504.2 | XP_011535806.1 | 3 | 954 | |||
XM_011537506.2 | XP_011535808.1 | 3 | 954 | |||
XM_011537507.2 | XP_011535809.1 | 3 | 954 | |||
XM_011537509.2 | XP_011535811.1 | 3 | 954 | |||
XM_017020629.1 | XP_016876118.1 | 3 | 954 | |||
XM_017020630.1 | XP_016876119.1 | 3 | 954 | |||
XM_017020631.1 | XP_016876120.1 | 3 | 954 | |||
XM_017020632.1 | XP_016876121.1 | 3 | 954 | |||
XM_017020633.1 | XP_016876122.1 | 3 | 954 | |||
XM_017020634.1 | XP_016876123.1 | 3 | 954 | |||
XM_017020635.1 | XP_016876124.1 | 3 | 954 | |||
XM_017020636.1 | XP_016876125.1 | 3 | 954 | |||
GenBank mRNA | AF209505.1 | - | 2 | 168 | ||
AK000630.1 | - | 3 | 851 | |||
AY358768.1 | - | 3 | 732 | |||
BC012564.1 | - | 3 | 916 | 4080970 | ||
BC068515.1 | - | 3 | 952 | 4819377 |
