siRNA Details
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018946.3 | NP_061819.2 | 5 | 755 | ||
XM_011518787.2 | XP_011517089.1 | 5 | 511 | |||
XM_011518788.2 | XP_011517090.1 | 3 | 262 | |||
XM_017014811.1 | XP_016870300.1 | 4 | 456 | |||
GenBank mRNA | AF161387.1 | - | 2 | 54 | ||
AF257466.1 | - | 5 | 623 | |||
AK001659.1 | - | 5 | 693 | |||
AK091184.1 | - | 3 | 2549 | |||
AK316608.1 | - | 5 | 715 | |||
AL526281.3 | - | 5 | 755 | |||
BC000008.2 | - | 5 | 701 | 3505324 | ||
BC019315.1 | - | 5 | 683 | 2821389 | ||
FJ224349.1 | - | 5 | 623 |

More Information
Other Species Targeted

Gene Ontology Categories:
