siRNA Details
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 2 | 203 | ||
NM_020746.4 | NP_065797.2 | 2 | 203 | |||
NR_037921.1 | 2 | 203 | ||||
GenBank mRNA | AB033097.1 | - | 1 | 109 | ||
AB097003.1 | - | 2 | 166 | |||
AB232371.1 | - | 1 | 103 | |||
AK023799.1 | - | 2 | 159 | |||
AK123956.1 | - | 2 | 164 | |||
AK130992.1 | - | 2 | 124 | |||
AK291785.1 | - | 2 | 162 | |||
AK296897.1 | - | 2 | 168 | |||
BC044952.1 | - | 2 | 137 | 5751684 | ||
BM834250.1 | - | 2 | 167 | |||
BX649084.1 | - | 2 | 164 | |||
DB219666.1 | - | 2 | 203 | |||
DQ167126.1 | - | 1 | 31 | |||
DQ174270.1 | - | 1 | 31 | |||
DQ181928.1 | - | 1 | 31 | |||
EF467323.1 | - | 1 | 31 | |||
EF467324.1 | - | 1 | 31 | |||
KC415005.1 | - | 1 | 31 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
