siRNA Details
Gene Symbol: DAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291963.1 | NP_001278892.1 | 3 | 592 | ||
NM_004394.2 | NP_004385.1 | 4 | 635 | |||
GenBank mRNA | AK298670.1 | - | 3 | 545 | ||
BC002726.2 | - | 4 | 592 | 3631886 | ||
BI823002.1 | - | 4 | 465 | |||
BT006925.1 | - | 3 | 301 | |||
X76105.1 | - | 4 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
