siRNA Details
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039001.1 | NP_001034090.1 | 13 | 1430 | ||
GenBank mRNA | BC083909.1 | - | 13 | 1430 | 7134204 |

More Information
Other Species Targeted

Gene Ontology Categories:
