siRNA Details
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039001.1 | NP_001034090.1 | 3, 4 | 362 | ||
GenBank mRNA | BC083909.1 | - | 3, 4 | 362 | 7134204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
