siRNA Details
Gene Symbol: C6orf226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008739.1 | NP_001008739.1 | 1 | 435 | ||
GenBank mRNA | BC051007.1 | - | 1 | 435 | ||
BC060325.1 | - | 1 | 423 | 4364495 | ||
BC073804.1 | - | 1 | 435 | 6254518 | ||
BC094854.1 | - | 1 | 435 | 6211808 |
