siRNA Details
Gene Symbol: FAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017178.1 | XP_016872667.1 | 26, 27 | 13254 | ||
XM_017017179.1 | XP_016872668.1 | 27, 28 | 13476 | |||
XM_017017181.1 | XP_016872670.1 | 25, 26 | 13184 | |||
XM_017017185.1 | XP_016872674.1 | 25, 26 | 11080 | |||
XM_017017186.1 | XP_016872675.1 | 25, 26 | 10368 | |||
XM_017017187.1 | XP_016872676.1 | 25, 26 | 14726 | |||
GenBank mRNA | AB076400.1 | - | 8, 9 | 2577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
