siRNA Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010844.3 | NP_001010844.1 | 3 | 485 | ||
XM_006715336.3 | XP_006715399.1 | 3 | 526 | |||
XM_011535447.1 | XP_011533749.1 | 3 | 525 | |||
XM_017010264.1 | XP_016865753.1 | 3 | 304 | |||
GenBank mRNA | AK098678.1 | - | 3 | 982 | ||
AK309003.1 | - | 3 | 422 | |||
BC112253.1 | - | 3 | 474 | 8327721 | ||
BC112255.1 | - | 3 | 474 | 8327723 | ||
BX103697.1 | - | 1 | 61 | |||
CR990080.1 | - | 3 | 485 |
