siRNA Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039188.1 | NP_001034277.1 | 7, 8 | 1151 | ||
NM_001177868.1 | NP_001171339.1 | 7, 8 | 876 | |||
NM_001177869.1 | NP_001171340.1 | 6, 7 | 812 | |||
NM_026830.2 | NP_081106.1 | 7, 8 | 1119 | |||
GenBank mRNA | AK047047.1 | - | 7, 8 | 2070 | ||
AK047251.1 | - | 7, 8 | 2072 | |||
AK147260.1 | - | 7, 8 | 2274 | |||
AK147653.1 | - | 7, 8 | 2274 | |||
AK154980.1 | - | 7, 8 | 1119 | |||
AK170652.1 | - | 7, 8 | 1151 | |||
AK171375.1 | - | 7, 8 | 856 | |||
AY946044.1 | - | 4, 5 | 503 | |||
BC080680.1 | - | 7, 8 | 965 | 6330949 |
