siRNA Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177869.1 | NP_001171340.1 | 12 | 7354 | ||
GenBank mRNA | AK004146.1 | - | 1 | 763 | ||
AK147260.1 | - | 12 | 8654 | |||
AK147653.1 | - | 12 | 8654 | |||
AY946044.1 | - | 10 | 7045 | |||
BC027834.1 | - | 2 | 2908 | |||
BC051573.1 | - | 1 | 364 |
