siRNA Details
Gene Symbol: Fam149b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024512.2 | NP_001019683.1 | 14 | 2254 | ||
NM_172379.3 | NP_759011.2 | 12 | 1975 | |||
GenBank mRNA | AK034594.1 | - | 1 | 3018 | ||
AK129252.1 | - | 11 | 1654 | |||
BC057907.1 | - | 12 | 1877 | 5064176 | ||
BC058650.1 | - | 13 | 3971 | |||
BC059221.1 | - | 13 | 3971 | |||
BC069846.1 | - | 14 | 2152 | 4196478 | ||
BC080849.1 | - | 14 | 2194 | 5717789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
