siRNA Details
Gene Symbol: Fam149b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024512.2 | NP_001019683.1 | 10 | 1376 | ||
NM_172379.3 | NP_759011.2 | 8 | 1097 | |||
GenBank mRNA | BC008267.1 | - | 4 | 440 | ||
BC057907.1 | - | 8 | 999 | 5064176 | ||
BC058650.1 | - | 9 | 3096 | |||
BC059221.1 | - | 9 | 3096 | |||
BC069846.1 | - | 10 | 1274 | 4196478 | ||
BC080849.1 | - | 10 | 1316 | 5717789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
