siRNA Details
Gene Symbol: 2810474O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 851 | ||
NM_001289662.1 | NP_001276591.1 | 3 | 764 | |||
NM_026054.3 | NP_080330.1 | 3 | 782 | |||
GenBank mRNA | AK013408.1 | - | 5 | 909 | ||
AK017376.1 | - | 3 | 768 | |||
AK031526.1 | - | 3 | 647 | |||
AK031797.1 | - | 5 | 868 | |||
AK031989.1 | - | 4 | 815 | |||
AK079405.1 | - | 4 | 680 | |||
AK220403.1 | - | 3 | 765 | |||
BC086679.1 | - | 1 | 204 | |||
BC139435.1 | - | 3 | 704 | 8862457 | ||
BC139436.1 | - | 4 | 791 | 8862458 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008763457, XM_006237708, XM_006237707.
Gene Ontology Categories:
