siRNA Details
Gene Symbol: 2810474O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 3120 | ||
NM_001289662.1 | NP_001276591.1 | 3 | 3033 | |||
NM_026054.3 | NP_080330.1 | 3 | 3051 | |||
GenBank mRNA | AK031526.1 | - | 3 | 2914 | ||
AK031989.1 | - | 4 | 3083 | |||
AK079405.1 | - | 4 | 2951 | |||
AK220403.1 | - | 3 | 3034 | |||
BC023168.1 | - | 1 | 1027 | |||
BC086679.1 | - | 1 | 2471 | |||
BC139435.1 | - | 3 | 2973 | 8862457 | ||
BC139436.1 | - | 4 | 3060 | 8862458 |
