siRNA Details
Gene Symbol: PLEKHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100623.1 | NP_001094093.1 | 7, 8 | 1100 | ||
NM_001267064.1 | NP_001253993.1 | 7, 8 | 1121 | |||
NM_001267065.1 | NP_001253994.1 | 7, 8 | 1124 | |||
NM_001267067.1 | NP_001253996.1 | 5, 6 | 970 | |||
NM_001267068.1 | NP_001253997.1 | 5, 6 | 967 | |||
NM_001309448.1 | NP_001296377.1 | 6, 7 | 1055 | |||
NM_001309450.1 | NP_001296379.1 | 5, 6 | 957 | |||
NM_001309451.1 | NP_001296380.1 | 6, 7 | 1052 | |||
NM_001309452.1 | NP_001296381.1 | 8, 9 | 1198 | |||
NM_017958.2 | NP_060428.2 | 7, 8 | 1097 | |||
XM_017004410.1 | XP_016859899.1 | 6, 7 | 592 | |||
XM_017004411.1 | XP_016859900.1 | 8, 9 | 1728 | |||
XM_017004412.1 | XP_016859901.1 | 8, 9 | 1704 | |||
GenBank mRNA | AK000790.1 | - | 7, 8 | 608 | ||
AK093730.1 | - | 7, 8 | 1335 | |||
AK123710.1 | - | 7, 8 | 1057 | |||
AK126289.1 | - | 7, 8 | 1124 | |||
AK223338.1 | - | 7, 8 | 608 | |||
AK296008.1 | - | 6, 7 | 564 | |||
AK302770.1 | - | 5, 6 | 512 | |||
BC013991.2 | - | 7, 8 | 593 | 3639839 | ||
BC050695.1 | - | 7, 8 | 595 | 6162960 | ||
BQ649140.1 | - | 7, 8 | 630 | |||
BU174451.1 | - | 8, 9 | 679 | |||
CK000843.1 | - | 5, 6 | 460 | |||
DC303684.1 | - | 5, 6 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
