siRNA Details
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 5 | 551 | ||
NM_001286221.1 | NP_001273150.1 | 7 | 682 | |||
NM_001286222.1 | NP_001273151.1 | 7 | 736 | |||
NM_001286223.1 | NP_001273152.1 | 5 | 532 | |||
NM_027696.3 | NP_081972.2 | 7 | 739 | |||
GenBank mRNA | AK129405.1 | - | 7 | 659 | ||
AK138876.1 | - | 5 | 552 | |||
AK143201.1 | - | 5 | 657 | |||
AK168693.1 | - | 5 | 532 | |||
AY769435.1 | - | 6 | 531 | |||
AY769436.1 | - | 5 | 454 | |||
AY769437.1 | - | 5 | 504 | |||
AY769438.1 | - | 7 | 604 | |||
BC043930.1 | - | 7 | 664 | |||
BC108273.1 | - | 5 | 518 | 30080243 | ||
BC130227.1 | - | 5 | 503 | 40141863 | ||
BQ562973.1 | - | 2 | 129 | |||
CN538186.1 | - | 2 | 152 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001131012, XM_008763881, XM_006238479, XM_008763879, XM_006238484, and others.
Gene Ontology Categories:
