siRNA Details
Gene Symbol: Inpp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290797.1 | NP_001277726.1 | 12, 13 | 1503 | ||
NM_001290798.1 | NP_001277727.1 | 2, 3 | 379 | |||
NM_001290799.1 | NP_001277728.1 | 2, 3 | 379 | |||
NM_030266.4 | NP_084542.2 | 12, 13 | 1503 | |||
NM_172971.3 | NP_766559.2 | 2, 3 | 379 | |||
GenBank mRNA | AF317838.1 | - | 10, 11 | 1056 | ||
AK035867.1 | - | 2, 3 | 380 | |||
AK149414.1 | - | 12, 13 | 1428 | |||
BC120541.1 | - | 2, 3 | 375 | 40129464 | ||
BC144747.1 | - | 2, 3 | 375 | |||
BC144748.1 | - | 2, 3 | 375 | 9053296 |
