siRNA Details
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 7 | 3257 | |||
GenBank mRNA | AL117401.1 | - | 7 | 3051 | ||
AL117484.1 | - | 4 | 1453 | |||
BC063879.1 | - | 2 | 754 | |||
BC086868.1 | - | 3 | 1579 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 3310 | |||
GenBank mRNA | BC051721.1 | - | 6 | 3310 |
Gene Symbol: LOC107987388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030130.1 | XP_016885619.1 | 4 | 2415 | ||
XM_017030131.1 | XP_016885620.1 | 6 | 1868 | |||
XM_017030132.1 | XP_016885621.1 | 5 | 1888 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 6 | 7509 | |||
GenBank mRNA | BX648116.1 | - | 5 | 2601 |
Gene Symbol: POM121L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024591.1 | 10 | 3029 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 4371 | |||
GenBank mRNA | AK292222.1 | - | 4 | 1255 | ||
AL117485.1 | - | 5 | 2322 | |||
BC050331.1 | - | 2 | 555 | |||
BC068032.1 | - | 2 | 416 | |||
BC090873.1 | - | 2 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
