siRNA Details
Gene Symbol: TMEM248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017994.4 | NP_060464.1 | 4 | 734 | ||
XM_005250482.3 | XP_005250539.1 | 4 | 586 | |||
GenBank mRNA | AK000961.1 | - | 4 | 720 | ||
AK222774.1 | - | 4 | 734 | |||
BC008675.1 | - | 4 | 714 | 3853834 | ||
BC010519.1 | - | 4 | 688 | 4150551 | ||
BC012562.1 | - | 4 | 729 | 4332094 | ||
BC041764.2 | - | 4 | 703 | 4152659 |
