siRNA Details
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206847.1 | NP_996729.1 | 22 | 2323 | ||
XM_008771885.1 | XP_008770107.1 | 22 | 2383 | |||
GenBank mRNA | BC088847.1 | - | 22 | 2323 | 7312293 | |
FN432820.1 | - | 22 | 2351 | |||
L25387.1 | - | 22 | 2297 |
