siRNA Details
Gene Symbol: Krt42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212483.2 | NP_997648.2 | 8 | 1436 | ||
GenBank mRNA | AK019115.1 | - | 5 | 953 | ||
AK019133.1 | - | 5 | 898 | |||
AY459292.1 | - | 8 | 1436 | |||
BC112371.1 | - | 8 | 1428 | |||
BC148204.1 | - | 8 | 1436 | 40056537 |
