siRNA Details
Gene Symbol: NPSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300933.1 | NP_001287862.1 | 8 | 1100 | ||
NM_001300934.1 | NP_001287863.1 | 6 | 935 | |||
NM_001300935.1 | NP_001287864.1 | 8 | 1133 | |||
NM_207172.1 | NP_997055.1 | 8 | 1087 | |||
NM_207173.1 | NP_997056.1 | 8 | 1087 | |||
GenBank mRNA | AY310326.1 | - | 8 | 1087 | ||
AY310327.1 | - | 8 | 1087 | |||
AY310328.1 | - | 8 | 1054 | |||
AY310330.1 | - | 7 | 983 | |||
AY310331.1 | - | 7 | 993 | |||
AY310332.1 | - | 6 | 889 | |||
AY956446.1 | - | 8 | 992 | |||
AY956447.1 | - | 8 | 963 | |||
AY956448.1 | - | 8 | 963 | |||
BC132693.1 | - | 8 | 1004 | 40146715 | ||
BC132695.1 | - | 8 | 1004 | 40146717 | ||
BC144380.1 | - | 6 | 806 | 9052901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
